Synonym: β-Naphthylamine, 2-Aminonaphthalene
CAS Number 91-59-8
Linear Formula C10H7NH2
Molecular Weight 143.19
Beilstein Registry Number 3939429
EC Number 202-080-4
MDL number MFCD00004112
PubChem Substance ID 24869746
| Related Categories | All alphabetical sorted, Analytical Standards, Analytical/Chromatography, Chromatography, Colors, |
| grade | analytical standard, for environmental analysis |
| bp | 306 °C(lit.) |
| mp | 111-113 °C(lit.) |
| density | 1.061 g/mL at 25 °C(lit.) |
Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?