31667 Fluka

Chloramphenicol

VETRANAL, analytical standard

DOWNLOAD MSDS (PDF)

Synonym: D-(−)-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, D-(−)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]acetamide, Chloromycetin

  • CAS Number 56-75-7

  • Linear Formula Cl2CHCONHCH(CH2OH)CH(OH)C6H4NO2

  • Molecular Weight 323.13

  •  Beilstein Registry Number 2225532

  •  EC Number 200-287-4

  •  MDL number MFCD00078159

  •  PubChem Substance ID 24869808

Properties

Related Categories Bioactive Small Molecules, C-CH, Cell Biology More...
grade   VETRANAL, analytical standard
mp   148-150 °C(lit.)
Gene Information   human ... CYP1A2(1544)

Description

Biochem/physiol Actions

Mode of Action: Inhibits translation on the 50S ribosomal subunit at the peptidyltransferase step (elongation inhibition). Bacteriostatic.
Mode of Resistance: Acetylation by chloramphenicol acetyltransferase (cat gene).

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706024

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Chloramphenicol

≥98% (TLC)

Chloramphenicol

γ-irradiated

Chloramphenicol

BioReagent, suitable for plant cell culture

Chloramphenicol

meets USP testing specifications

Chloramphenicol

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?