Synonym: Ammonia aqueous, Ammonia water
CAS Number 1336-21-6
Linear Formula NH4OH
Molecular Weight 35.05
Beilstein Registry Number 3587154
MDL number MFCD00066650
PubChem Substance ID 24859136
Popular Documents: Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Base Solutions, Titration, Volumetric Solutions More... |
| quality | volumetric |
| expl. lim. | 27 % |
| concentration | 4.90-5.10 N (Exact value stated on label) |
| 5.0 M NH4OH (5.0N) | |
| density | 0.9 g/mL at 25 °C(lit.) |
| 0.965 g/mL at 25 °C |
Visit our Titration Center to learn more.
2 L in poly bottle
500 mL in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?