• USA Home
  • 318949 - Hydrochloric acid solution

EMAIL THIS PAGE TO A FRIEND
318949 Fluka

Hydrochloric acid solution

volumetric, 1.0 M HCl (1.0N)

Synonym: Hydrogen chloride solution

Purchase

Properties

Related Categories Acid Solutions, Acids, Acids & Bases, Analytical Reagents, Analytical/Chromatography,
vapor pressure   190 mmHg ( 25 °C)
quality   volumetric
concentration   0.950-1.050 N (exact value stated on label)
  1.0 M HCl (1.0N)
bp   100 °C(lit.)
density   1.017 g/mL at 25 °C

Description

General description

Visit our Titration Center to learn more.

Packaging

500, 6×500 mL in poly bottle

6×1, 2 L in poly bottle

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
1
Protocols & Articles

Protocols

Enzymatic Assay of Pepsin (3.4.23.1)

This procedure may be used for determination of Pepsin activity using hemoglobin as the substrate. It is a spectrophotometric stop rate determination.

Enzymatic Assay of Proteinase K (EC 3.4.21.64)

This procedure may be used for determination of Proteinase K activity using hemoglobin as the substrate. It is not used to assay the activity of immobilized Proteinase K products, such as Catalog Num...
Keywords: Immobilization

Enzymatic Assay of Trypsin Inhibitor

This procedure applies to all products that have a specification for Trypsin Inhibition at Sigma-Aldrich, St. Louis.

Enzymatic Assay of Urease from Jack Beans

The scope of this procedure is for all products that have a specification for the enzymatic determination of Urease, from Jack Beans. This assay is not to be used to determine the activity of Urease,...
Keywords: Indicators, Sonication, Titrations

Procedure for Enzymatic Assay of Chymotrypsin (α‑Chymotrypsin, EC 3.4.21.1)

This procedure is for products with a specification for Chymotrypsin activity. It is not to be used to assay Insoluble Chymotrypsin such as Catalog No. C9134. The procedure is a continuous spectropho...
Keywords: Extinction coefficient

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?