• USA Home
  • 318965 - Hydrochloric acid solution

EMAIL THIS PAGE TO A FRIEND
318965 Fluka

Hydrochloric acid solution

volumetric, 0.1 M HCl (0.1N)

Synonym: Hydrogen chloride solution

Purchase

Properties

Related Categories Acid Solutions, Analytical Reagents, Analytical/Chromatography, Hydrochloric acid solution, Most common compounds for volumetric titration,
vapor pressure   190 mmHg ( 25 °C)
quality   volumetric
concentration   0.0950-0.1050 N (exact value stated on label)
  0.1 M HCl (0.1N)
bp   100 °C(lit.)
density   1 g/mL at 25 °C

Description

Analysis Note

Traceable to NIST SRM 723

General description

Visit our Titration Center to learn more.

Packaging

2, 4×4 L in poly bottle

500 mL in poly bottle

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?