• USA Home
  • 319317 - Potassium dichromate solution

EMAIL THIS PAGE TO A FRIEND
319317 Fluka

Potassium dichromate solution

volumetric, 1/60 M K2Cr2O7 (0.1N)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Chromates, Most common compounds for volumetric titration,
quality   volumetric
concentration   0.0950-0.1050 N (Exact value stated on label)
  1/60 M K2Cr2O7 (0.1N)
density   1.000 g/mL at 25 °C

Description

General description

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?