• USA Home
  • 319430 - Silver nitrate solution

EMAIL THIS PAGE TO A FRIEND
319430 Fluka

Silver nitrate solution

volumetric, 0.1 M AgNO3 (0.1N)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Most common compounds for volumetric titration, S - Z, Salt Solutions,
vapor density   5.8 (vs air)
quality   volumetric
concentration   0.0950-0.1050 N (Exact value stated on label/CoA)
  0.1 M AgNO3 (0.1N)
density   1.015 g/mL at 25 °C

Description

General description

Visit our Titration Center to learn more.

Packaging

2 L in glass bottle

500 mL in glass bottle

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?