• USA Home
  • 319503 - Sodium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
319503 Fluka

Sodium hydroxide solution

volumetric, 0.5 M NaOH (0.5N)

Purchase

Properties

Related Categories Acids & Bases, Analytical Reagents, Analytical/Chromatography, Base Solutions, Bases,
vapor density   >1 (vs air)
vapor pressure   3 mmHg ( 37 °C)
quality   volumetric
concentration   0.4900-0.5100 N (Exact value stated on label)
  0.5 M NaOH (0.5N)
density   1.02 g/mL at 25 °C

Description

General description

Visit our Titration Center to learn more.

Packaging

2 L in poly bottle

500 mL in poly bottle

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1824 8 / PGIII
WGK Germany 
nwg
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?