CAS Number 7772-98-7
Linear Formula Na2S2O3
Molecular Weight 158.11
EC Number 231-791-2
MDL number MFCD00003499
PubChem Substance ID 24859174
Popular Documents: Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Most common compounds for volumetric titration, S - Z, Salt Solutions, |
| quality | volumetric |
| concentration | 0.0950-0.1050 N (Exact value stated on label) |
| 0.1 M Na2S2O3 (0.1N) | |
| density | 1.01 g/mL at 25 °C |
Visit our Titration Center to learn more.
2 L in poly bottle
500 mL in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?