31999 Fluka

Sodium 5,5-diethylbarbiturate

BioUltra, ≥99.5% (NT)

DOWNLOAD MSDS (PDF)

Synonym: 5,5-Diethylbarbituric acid sodium salt, Barbitone, Sodium barbital, Veronal sodium salt

  • CAS Number 144-02-5

  • Empirical Formula (Hill Notation) C8H11N2NaO3

  • Molecular Weight 206.17

  •  Beilstein Registry Number 3921202

  •  EC Number 205-613-9

  •  MDL number MFCD00036203

Properties

Related Categories Alphabetical, BioUltra Buffers, Biological Buffers More...
product line   BioUltra
assay   ≥99.5% (NT)
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
impurities   insoluble matter, passes filter test
pH   9.0-10.5 (25 °C, 0.5 M in H2O)
solubility   H2O: soluble0.5 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤10 mg/kg
  Ca: ≤20 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤500 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
abs.   absorption0.5M/290 nm, H2O cut-off

Description

Biochem/physiol Actions

Sedative/hypnotic

Other Notes

Buffer component for the separation of serum proteins by cellulose acetate electrophoresis1

Sales restrictions may apply

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?