Synonym:
5,5-
CAS Number 144-02-5
Empirical Formula (Hill Notation) C8H11N2NaO3
Molecular Weight 206.17
Beilstein Registry Number 3921202
EC Number 205-613-9
MDL number MFCD00036203
| grade | purum |
| assay | ≥99.0% (NT) |
| drug control | regulated under CDSA - not available from Sigma-Aldrich Canada |
Sedative/hypnotic
Sales restrictions may apply
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?