• USA Home
  • 320145 - Ammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
320145 Fluka

Ammonium hydroxide solution

ACS reagent, 28.0-30.0% NH3 basis

Synonym: Ammonia aqueous, Ammonia water

  • CAS Number 1336-21-6

  • Linear Formula NH4OH

  • Molecular Weight 35.05

  •  Beilstein Registry Number 3587154

  •  MDL number MFCD00066650

  •  PubChem Substance ID 24859252

  •  eCl@ss 38060103

Purchase

Properties

grade   ACS reagent
vapor density   1.2 (vs air)
vapor pressure   115 mmHg ( 20 °C)
assay   28.0-30.0% NH3 basis
expl. lim.   27 %
impurities   MnO4- reducers, passes test
  ≤0.002% CO2
ign. residue   ≤0.002%
density   0.9 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤0.5 ppm
  nitrate (NO3-): ≤2 ppm
  phosphate (PO43-): ≤2 ppm
  sulfate (SO42-): ≤2 ppm
cation traces   Fe: ≤0.2 ppm
  heavy metals (as Pb): ≤0.5 ppm

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2672 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

related product

Product #

Description

Add to Cart

Z568724 QianCap safety cap system for Ace pressure tubes and bottles, QianCap-15, fits no. 15 internal thread

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?