Synonym:
2,3-
CAS Number 1563-66-2
Empirical Formula (Hill Notation) C12H15NO3
Molecular Weight 221.25
Beilstein Registry Number 1428746
EC Number 216-353-0
MDL number MFCD00041819
PubChem Substance ID 24868844
| Related Categories | Acaricides, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, C, |
| grade | PESTANAL®, analytical standard |
| mp | 150-153 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?