Synonym:
2,4-
CAS Number 122-34-9
Empirical Formula (Hill Notation) C7H12ClN5
Molecular Weight 201.66
Beilstein Registry Number 10895
EC Number 204-535-2
MDL number MFCD00023174
PubChem Substance ID 24869206
| Related Categories | 2000/60/EC, Algicides, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?