• USA Home
  • 32085 - Diethyl chlorophosphate

EMAIL THIS PAGE TO A FRIEND

32085 Fluka

Diethyl chlorophosphate

purum, >97.0% (T)

Synonym: Diethyl phosphorochloridate

Purchase

Properties

vapor density   5.94 (vs air)
vapor pressure   0.1 mmHg ( 25 °C)
grade   purum
assay   >97.0% (T)
refractive index   n20/D 1.416(lit.)
bp   60 °C/2 mmHg(lit.)
density   1.194 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by D91632-ALDRICH | Diethyl chlorophosphate 97%

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
T+
Risk Statements (Europe) 
Safety Statements (Europe) 
28-36/37/39-45
RIDADR 
UN 3278 6.1 / PGI
WGK Germany 
3
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 1,IV,1345

Fieser 1,248 / Fieser 3,98 / Fieser 4,152 / Fieser 5,217 / Fieser 6,192 / Fieser 8,171

Aldrich MSDS 1, 664:D / Corp MSDS 1 (1), 1201:D / FT-IR 1 (1), 922:B / FT-IR 2 (1), 1579:C / FT-NMR 1 (1), 1498:A / IR-Spectra (3), 554:B / IR-Spectra (2), 491:B / NMR-Reference 2 (2), 872:C / RegBook 1 (1), 1117:B / Sax 6, 1007 / Sigma FT-IR 1 (2), 459:C / Structure Index 1, 173:D:1 / Vapor Phase 3, 844:B

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?