32477 Fluka

Moxifloxacin hydrochloride

VETRANAL, analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1-Cyclopropyl-6-fluoro-8-methoxy-7-[(4aS,7aS)-octahydropyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

  • CAS Number 186826-86-8

  • Empirical Formula (Hill Notation) C21H24FN3O4· HCl

  • Molecular Weight 437.89

  •  Beilstein Registry Number 8377447

  •  MDL number MFCD00949117

Properties

grade   VETRANAL, analytical standard

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Ciprofloxacin

≥98.0% (HPLC)

Levofloxacin

≥98.0% (HPLC)

Linezolid
PZ0014Linezolid

≥98% (HPLC)

fluoroquinolone antibiotic

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?