Synonym:
2-
CAS Number 103361-09-7
Empirical Formula (Hill Notation) C19H15FN2O4
Molecular Weight 354.33
MDL number MFCD01754256
| Related Categories | All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Chromatography, Cosmetic Standards, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?