Synonym:
1-Ethyl-d5-
CAS Number 1228182-51-1
Empirical Formula (Hill Notation) C17D5H15FN3O3
Molecular Weight 338.39
MDL number MFCD16652561
| Related Categories | Analytical Standards, Analytical/Chromatography, Antibiotics, Chemical Structure, Chromatography, |
| grade | VETRANAL™, analytical standard |
| mass shift | M+5 |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?