EMAIL THIS PAGE TO A FRIEND

32568 Fluka

Cimaterol

VETRANAL, analytical standard

Synonym: 2-Amino-5-(1-hydroxy-2-isopropylaminoethyl)benzonitrile

  • CAS Number 54239-37-1

  • Empirical Formula (Hill Notation) C12H17N3O

  • Molecular Weight 219.28

  •  Beilstein Registry Number 2418129

  •  MDL number MFCD00209815

Purchase

Properties

grade   analytical standard
product line   VETRANAL
form   neat
format   neat

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Brombuterol hydrochloride

VETRANAL, analytical standard

Cimaterol-d<SUB>7</SUB>

VETRANAL, analytical standard

Cimbuterol

VETRANAL, analytical standard

Mabuterol hydrochloride

VETRANAL, analytical standard

Mapenterol hydrochloride

VETRANAL, analytical standard

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37
WGK Germany 
3
RTECS 
DI2457930

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Effect of duration of a beta-agonist treatment on growth, feed intake and carcass characteristics in finishing bulls. Fiems LO, Boucque CV, and Cottyn BG Arch. Tierernahr. 45(2), 101-9, (1993)

Loading...


[Simultaneous determination of cimaterol, clenbuterol and salbutamol in feeds by capillary zone electrophoresis]. Duan J, Chen H, Chen Y, et al. Se Pu 23(3), 261-3, (2005)

Loading...


Long-term effects of cimaterol in Friesian steers: II. Carcass composition and meat quality. Chikhou FH, Moloney AP, Allen P, et al. J. Anim. Sci. 71(4), 914-22, (1993)

Loading...


Gas and liquid chromatography coupled to tandem mass spectrometry for the multiresidue analysis of beta-agonists in biological matrices. Van Vyncht G, Preece S, Gaspar P, et al. J. Chromatogr. A 750(1-2), 43-9, (1996)

Loading...


Long-term effects of cimaterol in Friesian steers: I. Growth, feed efficiency, and selected carcass traits. Chikhou FH, Moloney AP, Allen P, et al. J. Anim. Sci. 71(4), 906-13, (1993)

Loading...


Field-amplified on-line sample stacking for separation and determination of cimaterol, clenbuterol and salbutamol using capillary electrophoresis. Shi Y, Huang Y, Duan J, et al. J. Chromatogr. A 1125(1), 124-8, (2006)

Loading...


Changes in calpain and calpastatin mRNA induced by beta-adrenergic stimulation of bovine skeletal muscle. Parr T, Bardsley RG, Gilmour RS, et al. Eur. J. Biochem. 208(2), 333-9, (1992)

Loading...


The effect of cimaterol on muscle fiber characteristics, capillary supply, and metabolic potentials of longissimus and semitendinosus muscles from young Friesian bulls. Vestergaard M, Henckel P, Oksbjerg N, et al. J. Anim. Sci. 72(9), 2298-306, (1994)

Loading...


The beta-agonist cimaterol directly enhances chronic protein accretion in skeletal muscle. Byrem TM, Beermann DH, and Robinson TF J. Anim. Sci. 76(4), 988-98, (1998)

Loading...


Effect of dietary clenbuterol and cimaterol on muscle composition, beta-adrenergic and androgen receptor concentrations in broiler chickens. Schiavone A, Tarantola M, Perona G, et al. J. Anim. Physiol. Anim. Nutr. (Berl.) 88(3-4), 94-100, (2004)

Loading...


Response time of broiler chickens to cimaterol: meat tenderness, muscle composition fiber size, and carcass characteristics. Gwartney BL, Jones SJ, and Calkins CR J. Anim. Sci. 70(7), 2144-50, (1992)

Loading...


The effect of acute and chronic administration of the beta-agonist, cimaterol, on protein synthesis in ovine skin and muscle. Nash JE, Rocha HJ, Buchan V, et al. Br. J. Nutr. 71(4), 501-13, (1994)

Loading...


Determination of beta-agonist residues in bovine urine using liquid chromatography-tandem mass spectrometry. Dickson LC, MacNeil JD, Lee S, et al. J. AOAC Int. 88(1), 46-56, (2005)

Loading...


Influence of diet and beta-agonist administration on plasma concentrations of growth hormone and insulin-like growth factor-1 in young steers. Dawson JM, Craigon J, Buttery PJ, et al. Br. J. Nutr. 70(1), 93-102, (1993)

Loading...


Steroid and beta-adrenergic receptor modifications in target organs of broiler chickens fed with a diet containing beta2-adrenergic agents. Badino P, Odore R, Pagliasso S, et al. Food Chem. Toxicol. 46(6), 2239-43, (2008)

Loading...


Effects of a beta 2-adrenergic agonist, cimaterol and corticosterone on growth and carcass composition of male rats. Brown J, Clasper C, Smith T, et al. Comp. Biochem. Physiol. Comp. Physiol. 102(1), 217-20, (1992)

Loading...


Determination of clenbuterol, salbutamol, and cimaterol in bovine retina by electrospray ionization-liquid chromatography-tandem mass spectrometry. Lau JH, Khoo CS, and Murby JE J. AOAC Int. 87(1), 31-8, (2004)

Loading...


Analysis and pharmacokinetics of cimaterol in growing Holstein steers. Byrem TM, Robinson TF, Boisclair YR, et al. J. Anim. Sci. 70(12), 3812-9, (1992)

Loading...


Effects of the beta-adrenergic agonist cimaterol on chicken muscle atrophy induced by stretch-release. Lee YB, Hitchcock L, and Summers PJ Growth. Dev. Aging 58(1), 13-9, (1994)

Loading...


Anticatabolic effect of the beta 2-agonist cimaterol in vivo in tumor-bearing animals. Stallion A, Foley-Nelson T, Chance WT, et al. J. Surg. Res. 59(3), 387-92, (1995)

Loading...


Effects of alpha2-adrenoceptor antagonists on metabolic processes of swine: II. Nitrogen balance responses. Cleale RM, Ingling JM, Search DJ, et al. J. Anim. Sci. 76(7), 1849-58, (1998)

Loading...


Characterization of dose-dependent metabolic responses to close arterial infusion of cimaterol in the hindlimb of steers. Byrem TM, Beermann DH, and Robinson TF J. Anim. Sci. 74(12), 2907-16, (1996)

Loading...


Anabolic impact of cimaterol in conjunction with enteral nutrition following burn trauma. Nelson JL, Chalk CL, and Warden GD J. Trauma 38(2), 237-41, (1995)

Loading...


Transient changes in growth and in calpain and calpastatin expression in ovine skeletal muscle after short-term dietary inclusion of cimaterol. Speck PA, Collingwood KM, Bardsley RG, et al. Biochimie 75(10), 917-23, (1993)

Loading...


Effects of increased beta 2-agonist dose in tumor-bearing animals. Stallion A, Foley-Nelson T, Chance WT, et al. Nutr. Cancer 20(3), 251-60, (1993)

Loading...


Merck 14,2278

Related Products

related product

Product #

Description

Add to Cart

32569 Cimaterol-d7, VETRANAL, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?