-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: Dimethyldiallylammonium chloride
CAS Number 7398-69-8
Linear Formula [(CH3)2N+(CH2CH=CH2)2]Cl-
Molecular Weight 161.67
Beilstein Registry Number 5830817
MDL number MFCD00043200
PubChem Substance ID 24859553
| Related Categories | Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents |
| grade | purum |
| concentration | ~65% in H2O (AT) |
| total impurities | ~3% polymeric components |
| refractive index |
n |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?