EMAIL THIS PAGE TO A FRIEND

32671 Fluka

Carnidazole

VETRANAL, analytical standard

Synonym: [2-(2-Methyl-5-nitro-imidazol-1-yl)ethyl]thiocarbamic acid O-methyl ester

  • CAS Number 42116-76-7

  • Empirical Formula (Hill Notation) C8H12N4O3S

  • Molecular Weight 244.27

  •  Beilstein Registry Number 620260

  •  EC Number 255-663-0

  •  MDL number MFCD00866619

Purchase

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Ipronidazole-OH

VETRANAL, analytical standard

Ornidazole

analytical standard

Secnidazole hemihydrate

VETRANAL, analytical standard

Ternidazole hydrochloride

VETRANAL, analytical standard

Tinidazole

VETRANAL, analytical standard

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
29-36-37-39
WGK Germany 
3
RTECS 
FD3912000

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?