EMAIL THIS PAGE TO A FRIEND

327719 Fluka

Polystyrene

analytical standard, for GPC, 2,500

Synonym: PS

Purchase

Properties

Related Categories Materials Science, Polymer Molecular Weight Standards, Reference/Calibration Standards More...
grade   analytical standard
  for GPC
form   neat
  powder
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   average Mw 2,500 (Typical)

Description

Packaging

1 g in glass bottle

Price and Availability

Customers Also Viewed

Polystyrene

analytical standard, for GPC, 800

Polystyrene

analytical standard, for GPC, 3,000

Polystyrene

analytical standard, for GPC, 4,000

Polystyrene

analytical standard, for GPC, 4,000

Polystyrene

analytical standard, for GPC, 13,000

Safety & Documentation

Safety Information

WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Aldrich MSDS 1, 1524:C / Corp MSDS 1 (2), 2886:D / FT-IR 1 (2), 1204:B / FT-IR 2 (3), 4676:A / FT-IR 2 (3), 4676:D / FT-IR 2 (3), 4677:A / FT-IR 2 (3), 4677:B / FT-IR 2 (3), 4675:A / FT-IR 2 (3), 4676:C / FT-IR 2 (3), 4674:D / FT-IR 2 (3), 4677:C / FT-IR 2 (3), 4675:D / FT-IR 2 (3), 4675:B / FT-IR 2 (3), 4676:B / FT-IR 1 (2), 1204:C / IR-Spectra (2), 1355:B / IR-Spectra (3), 1593:F / RegBook 1 (2), 3175:B / Sax 6, 2474 / Structure Index 1, 505:A:2

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?