Synonym:
2-(4-Chlorophenyl)-N-
CAS Number 374726-62-2
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Amide structure, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?