• USA Home
  • 32940 - Methyl Orange mixed indicator solution

EMAIL THIS PAGE TO A FRIEND
32940 Fluka

Methyl Orange mixed indicator solution

acid-base indicator

Synonym: Methyl orange – bromocresol green solution

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Indicator Solutions, Indicators, Titration More...
grade   acid-base indicator
visual transition interval   3.0 - 4.4
density   ≥0.999 g/mL at 20 °C

Description

General description

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?