Synonym:
1-
Empirical Formula (Hill Notation) C19D5H17FN3O3 · HCl
Molecular Weight 400.89
| grade | VETRANAL™, analytical standard |
| mass shift | M+5 |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?