EMAIL THIS PAGE TO A FRIEND
33129 Fluka

Dimethyl yellow

indicator

Synonym: 4-(Dimethylamino)azobenzene, N,N-Dimethyl-4-(phenylazo)aniline, Butter yellow, Methyl yellow

  • CAS Number 60-11-7

  • Linear Formula C6H5N=NC6H4N(CH3)2

  • Molecular Weight 225.29

  •  Beilstein Registry Number 746016

  •  Colour Index Number 11020

  •  EC Number 200-455-7

  •  MDL number MFCD00008308

  •  PubChem Substance ID 24859947

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Indicators, Indicators for non-aqueous titration, Non-aqueous Titration Reagents,
grade   indicator
visual transition interval   2.9 - 4.0, red to yellow
mp   111 °C (dec.)(lit.)
suitability   complies for TLC
  complies for change

Description

General description

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
BX7350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?