EMAIL THIS PAGE TO A FRIEND

33347 Fluka

AOZ

VETRANAL, analytical standard

Synonym: 3-Amino-2-oxazolidinone, NSC 111187, NSC 196570, NSC 38250

  • CAS Number 80-65-9

  • Empirical Formula (Hill Notation) C3H6N2O2

  • Molecular Weight 102.09

  •  Beilstein Registry Number 109430

  •  EC Number 201-299-2

  •  MDL number MFCD00020871

  •  PubChem Substance ID 24860134

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Chemical Structure,
grade   analytical standard
product line   VETRANAL
form   neat
mp   65-67 °C
suitability   passes test for identity

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

1-Aminohydantoin hydrochloride

VETRANAL, analytical standard

2-NP-AHD

VETRANAL, analytical standard

AMOZ
33349AMOZ

VETRANAL, analytical standard

AMOZ-d<SUB>5</SUB>
33881AMOZ-d5

VETRANAL, analytical standard

AOZ-d<SUB>4</SUB>
33880AOZ-d4

VETRANAL, analytical standard

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

related product

Product #

Description

Add to Cart

33868 2-NP-AOZ, VETRANAL, analytical standard
33880 AOZ-d4, VETRANAL, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?