Synonym:
4,4′-MDI,
4,4′-Methylenebis(phenyl isocyanate),
Bis(4-
CAS Number 101-68-8
Linear Formula CH2(C6H4NCO)2
Molecular Weight 250.25
Beilstein Registry Number 797662
EC Number 202-966-0
MDL number MFCD00036131
PubChem Substance ID 24860077
| Related Categories | Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard, for environmental analysis |
| bp | 200 °C/5 mmHg(lit.) |
| mp | 42-45 °C(lit.) |
| density | 1.18 g/mL at 25 °C(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?