Synonym: Doxycycline hydrochloride hemiethanolate hemihydrate
CAS Number 24390-14-5
Empirical Formula (Hill Notation) C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
Molecular Weight 512.94
Beilstein Registry Number 5702728
MDL number MFCD07357237
PubChem Substance ID 24860078
| Related Categories | 1694 Pharmaceuticals & Personal Care Products, A - K, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | VETRANAL™, analytical standard |
| suitability | suitable for 1694 per US EPA |
Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi1. It is a broad spectrum inhibitor of matrix metalloproteinases in vivo.
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?