Synonym: Clavulanate potassium
CAS Number 61177-45-5
Empirical Formula (Hill Notation) C8H8NO5K
Molecular Weight 237.25
EC Number 262-640-9
MDL number MFCD01710901
PubChem Substance ID 24860237
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, |
| grade | VETRANAL™, analytical standard |
| storage temp. | 2-8°C |
β-lactamase inhibitor, typically added to amoxicillin to increase its effectiveness.
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?