Synonym:
2,3,4,6,7,8,9,10-
CAS Number 6674-22-2
Empirical Formula (Hill Notation) C9H16N2
Molecular Weight 152.24
Beilstein Registry Number 508906
EC Number 229-713-7
MDL number MFCD00006930
PubChem Substance ID 24860258
Popular Documents: FTNMR (PDF)
| Related Categories | Chemical Synthesis, Organic Bases, Synthetic Reagents More... |
| grade | purum |
| assay | ≥98.0% (GC) |
| refractive index |
n |
| bp | 115 °C/11 mmHg(lit.) |
| density | 1.019 g/mL at 20 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?