33483 Fluka

4,4′-Diazido-2,2′-stilbenedisulfonic acid disodium salt tetrahydrate

≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: Disodium 4,4′-diazidostilbene-2,2′-disulfonate tetrahydrate

  • CAS Number 2718-90-3

  • Empirical Formula (Hill Notation) C14H8N6Na2O6S2 · 4H2O

  • Molecular Weight 538.42

  •  EC Number 220-316-4

  •  PubChem Substance ID 24860260

Description

Other Notes

A reagent for enzyme immobilisation on fibroin by photo-crosslinking. The reagent is water-soluble over a wide pH range1

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?