Synonym:
4,4′-Diisocyanato-methylenedicyclohexane,
4,4′-HMDI,
4,4′-Methylene-bis(cyclohexyl isocyanate),
Bis(4-
CAS Number 5124-30-1
Beilstein Registry Number 2217800
EC Number 225-863-2
MDL number MFCD00015405
PubChem Substance ID 24860266
| Related Categories | Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, |
| grade | analytical standard, for environmental analysis |
| quality | mixture of isomers |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?