CAS Number 91296-87-6 (anhydrous)
Empirical Formula (Hill Notation) C20H17F2N3O3 · HCl · xH2O
Molecular Weight 421.83 (anhydrous basis)
PubChem Substance ID 24860273
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, S, SA - SM More... |
| grade | VETRANAL™, analytical standard |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?