33552 Fluka

Buffer solution pH 13.0 (20 °C)

glycine / sodium hydroxide / sodium chloride solution

DOWNLOAD MSDS (PDF)
  •  EC Number 231-791-2

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Buffer Solutions, Buffer Solutions, pH 10 - 13, Titration,
pH   11.20 (90 °C)
  11.40 (80 °C)
  11.61 (70 °C)
  11.84 (60 °C)
  12.10 (50 °C)
  12.37 (40 °C)
  12.66 (30 °C)
  13.00 (20 °C)
  13.35 (10 °C)
  13.71 (0 °C)

Description

General description

This standard solution is tested against certified reference materials from NIST and/or PTB.

Visit our Titration Center to learn more.

Price and Availability

Customers Also Viewed

for 500 ml buffer solution, glycine / sodium hydroxide / sodium chloride solution

hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB

borax / sodium hydroxide

boric acid / sodium hydroxide solution / potassium chloride

di-sodium hydrogen phosphate / sodium hydroxide solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?