Synonym: Sodium tetraborate buffer solution
MDL number MFCD00081185
PubChem Substance ID 24860408
| Related Categories | Analytical Reagents, Analytical Standards, Analytical/Chromatography, Biological Buffers, Buffer Convenience Packaging, |
| quality | blue colored |
| pH | 8.64 (90 °C) |
| 8.67 (80 °C) | |
| 8.71 (70 °C) | |
| 8.75 (60 °C) | |
| 8.80 (50 °C) | |
| 8.86 (40 °C) |
This standard solution is tested against certified reference materials from NIST and/or PTB.
Visit our Titration Center to learn more.
The suffix -VP or -VOLPAC in the product number indicates packaging in 5 or 10 L VOLPAC containers.
Customers Also Viewed
violet colored, borax / sodium hydroxide
red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide
green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?