PubChem Substance ID 24860409
| Related Categories | Analytical Reagents, Analytical Standards, Analytical/Chromatography, Buffer Solutions, Buffer Standard Solutions, color-coded, |
| quality | violet colored |
| pH | 10.00 (20 °C) |
| 10.10 (10 °C) | |
| 10.24 (0 °C) | |
| 9.49 (90 °C) | |
| 9.55 (80 °C) | |
| 9.62 (70 °C) |
This standard solution is tested against certified reference materials from NIST and/or PTB.
Visit our Titration Center to learn more.
The suffix -VP or -VOLPAC in the product number indicates packaging in 5 or 10 L VOLPAC containers.
Customers Also Viewed
borax / sodium hydroxide
with fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST
red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide
potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide, traceable to SRM from NIST
green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?