Synonym:
(E)-N-
CAS Number 135410-20-7
Empirical Formula (Hill Notation) C10H11ClN4
Molecular Weight 222.67
MDL number MFCD06201842
| Related Categories | A, AA to AL, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| mp | 100-102 °C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?