Synonym: Baytril
CAS Number 93106-60-6
Empirical Formula (Hill Notation) C19H22FN3O3
Molecular Weight 359.39
Beilstein Registry Number 5307824
MDL number MFCD00792463
PubChem Substance ID 24860446
| Related Categories | 1694 Pharmaceuticals & Personal Care Products, A - K, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | VETRANAL™, analytical standard |
| suitability | suitable for 1694 per US EPA |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?