Synonym: Advocin®
CAS Number 112398-08-0
Empirical Formula (Hill Notation) C19H20FN3O3
Molecular Weight 357.38
MDL number MFCD00864910
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | VETRANAL™, analytical standard |
Advocin is a registered trademark of Pfizer, Inc.
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
VETRANAL™, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?