Synonym: 2-Chloro-N-(4′-chlorobiphenyl-2- yl)-nicotinamide
CAS Number 188425-85-6
Empirical Formula (Hill Notation) C18H12Cl2N2O
Molecular Weight 343.21
MDL number MFCD06795150
PubChem Substance ID 24860570
| Related Categories | Alphabetical Index of Analytical Standards, Amide structure, Analytical Standards, Analytical/Chromatography, B, |
| grade | PESTANAL®, analytical standard |
| form | solid |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?