33899 Fluka

Norfloxacin

VETRANAL, analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid

  • CAS Number 70458-96-7

  • Empirical Formula (Hill Notation) C16H18FN3O3

  • Molecular Weight 319.33

  •  EC Number 274-614-4

  •  MDL number MFCD00079532

  •  PubChem Substance ID 24860599

Properties

Related Categories 1694 Pharmaceuticals & Personal Care Products, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules,
grade   VETRANAL, analytical standard
suitability   suitable for 1694 per US EPA
storage temp.   2-8°C
Gene Information   human ... CYP1A2(1544)
rat ... Gabra1(29705)

Description

Biochem/physiol Actions

Norfloxacin blocks DNA replication by interfering with an ATP-induced structural transition of DNA complexed with DNA gyrase (topoisomerase).
Mode of action: inhibits bacterial DNA replication
Antimicrobial spectrum: Gram-negative bacteria; less effective against Gram-positive bacteria

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Ciprofloxacin

≥98.0% (HPLC)

Danofloxacin

VETRANAL, analytical standard

Enrofloxacin

VETRANAL, analytical standard

Norfloxacin

≥98% (TLC)

Norfloxacin-d<SUB>5</SUB>

VETRANAL, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?