Synonym:
1-
CAS Number 63451-35-4
Empirical Formula (Hill Notation) C20H11N2Na3O11S3
Molecular Weight 620.47
EC Number 264-197-7
MDL number MFCD00036393
PubChem Substance ID 24860637
| Related Categories | Analytical Reagents, Analytical/Chromatography, Indicators, Metal Titration Indicators, Titration More... |
| grade | indicator for metal titration |
| suitability | complies for suitability for complexometry |
Visit our Titration Center to learn more.
Customers Also Viewed

Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?