Synonym:
9-
CAS Number 115550-35-1
Empirical Formula (Hill Notation) C17H19FN4O4
Molecular Weight 362.36
MDL number MFCD00864820
PubChem Substance ID 24860807
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, M, MA - MD More... |
| grade | VETRANAL™, analytical standard |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Zeniquin is a registered trademark of Pfizer, Inc.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?