Synonym:
2,6-Dichloro-α-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)
CAS Number 101831-37-2
Empirical Formula (Hill Notation) C17H9Cl3N4O2
Molecular Weight 407.64
MDL number MFCD00867203
PubChem Substance ID 24860829
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, D, DIA - DIC More... |
| grade | VETRANAL™, analytical standard |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?