34139 Fluka

Fumonisin B1 solution

50 μg/mL in acetonitrile: water, analytical standard, for environmental analysis

DOWNLOAD MSDS (PDF)

Synonym: FB1

  • CAS Number 116355-83-0

  • Empirical Formula (Hill Notation) C34H59NO15

  • Molecular Weight 721.83

  •  EC Number 200-835-2

  •  PubChem Substance ID 24860938

  • Popular Documents:  Datasheet (PDF)  

Properties

Related Categories All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biotoxins,
grade   analytical standard, for environmental analysis
concentration   50 μg/mL in acetonitrile: water
storage temp.   −20°C

Description

General description

Certan Vial

Price and Availability

Customers Also Viewed

Fumonisin B<SUB>1</SUB>

reference material

Fumonisin B<SUB>2</SUB> from <I>Fusarium moniliforme</I>

sphingosine N-acyltransferase inhibitor

Fumonisin B<SUB>2</SUB> solution

50 μg/mL in acetonitrile: water (50:50), analytical standard, for environmental analysis

50 μg/mL in acetonitrile: water (each of FB1 and FB2), analytical standard, for environmental analysis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?