EMAIL THIS PAGE TO A FRIEND
34256 Fluka

Acetic acid solution

volumetric, 1.0 M CH3COOH (1.0N)

  • CAS Number 64-19-7

  • Linear Formula CH3CO2H

  • Molecular Weight 60.05

  •  Beilstein Registry Number 506007

  •  MDL number MFCD00036152

  •  PubChem Substance ID 24860710

Purchase

Description

General description

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?