Synonym:
(Ethylenedinitrilo)
CAS Number 139-33-3
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
Molecular Weight 336.21
Beilstein Registry Number 3822669
EC Number 231-791-2
MDL number MFCD00070672
PubChem Substance ID 24861328
Popular Documents: FTNMR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, By Reference Material, Complexometric Solutions (IDRANAL), Complexometric Titration Reagents (EDTA Titration), |
| grade | Reag. Ph. Eur. |
| for complexometry | |
| quality | volumetric |
| concentration | 0.1 M EDTA-Na2 (0.2N) |
Visit our Titration Center to learn more.
The suffix -VP or -VOLPAC in the product number indicates packaging in 5 or 10 L VOLPAC containers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?