EC Number 200-835-2
MDL number MFCD00001878
PubChem Substance ID 24861426
| Related Categories | Acetonitrile and Acetonitrile Solutions, Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, Blends- CHROMASOLV LC-MS, |
| grade | LC-MS CHROMASOLV® |
| assay | ≥99.5% (GC) |
| impurities | 0.093-0.107% formic acid |
| transmittance | 210 nm, ≥5% |
| 230 nm, ≥15% | |
| 254 nm, ≥90% | |
| cation traces | Ca: ≤0.5 mg/kg |
2.5 L in glass bottle
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
LC-MS CHROMASOLV®
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?