34672 Fluka

Methanol with 0.1% acetic acid

LC-MS CHROMASOLV®

DOWNLOAD MSDS (PDF)
  • Empirical Formula (Hill Notation) CH4O

  • Molecular Weight 32.04

  •  EC Number 200-659-6

  •  MDL number MFCD00004595

  •  PubChem Substance ID 24861432

Properties

Related Categories Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, Blends- CHROMASOLV LC-MS, Chromatography Reagents & Solvents,
grade   LC-MS CHROMASOLV®
assay   ≥99.5% (GC)
transmittance   210 nm, ≥5%
  230 nm, ≥50%
  254 nm, ≥95%
cation traces   Ca: ≤0.5 mg/kg
  K: ≤0.5 mg/kg
  Mg: ≤0.5 mg/kg
  Na: ≤2 mg/kg
abs.   HPLC-gradient/254 nm ≤10 mAU
suitability   LC-MS tested

Description

Packaging

2.5 L in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Acetonitrile with 0.1% formic acid

LC-MS CHROMASOLV®

Methanol

CHROMASOLV®, for HPLC, ≥99.9%

Methanol

LC-MS CHROMASOLV®

LC-MS CHROMASOLV®

Water with 0.1% acetic acid

LC-MS CHROMASOLV®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?